missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexanemethylamine, 97%, Thermo Scientific Chemicals

Catalog Number p-3770875
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
3218-02-8
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
3218-02-8

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 0.8700g/mL
Boiling Point 161°C to 163°C
Flash Point 43°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C7H15N
Linear Formula C6H11CH2NH2
Refractive Index 1.463
MDL Number MFCD00001520
Quantity 5 g
Beilstein 12, 12
Specific Gravity 0.87
Synonym cyclohexanemethylamine, aminomethylcyclohexane, aminomethyl cyclohexane, cyclohexanemethanamine, 1-cyclohexylmethanamine, hexahydrobenzylamine, 1-cyclohexylmethylamine, 4-aminomethylcyclohexane, c-cyclohexyl-methylamine, chembl1049
InChI Key AVKNGPAMCBSNSO-UHFFFAOYSA-N
SMILES C1CCC(CC1)CN
IUPAC Name cyclohexylmethanamine
Molecular Weight (g/mol) 113.2
PubChem CID 76688
Formula Weight 113.2
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Cyclohexanemethylamine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'