missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexane, Certified AR for Analysis, Fisher Chemical™

C6H12, CAS Number-110-82-7, 2.5L, 81 deg.C, CHEBI:29005, Colorless, 84.15g/mol, XDTMQSROBMDMFD-UHFFFAOYSA-N, cyclohexane, 6.5 deg.C, 3814, 84.162, Amber glass bottle, Liquid, 8078, C1CCCCC1, 2.9, 104mbar at 20 deg.C, 0.94 mPaS at 20 deg.C

Brand:  Fisher Chemical C/8921/25

677.00 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.



Code : 22

Additional Details : CAS Number : 110-82-7 Weight : 19.25000kg Transport : UN number : 1145 Chem class : 3 Pack group : II


 View more versions of this product

Product Code. 10151850

  • £794.00 / 25 L
Estimated Shipment: 08-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


For a copy of the TSE/BSE statement please contact Technical Support on Tel: 01509 555888 or email our support team for further assistance
 

Description

Description

C6H12, CAS Number-110-82-7, 2.5L, 81 deg.C, CHEBI:29005, Colorless, 84.15g/mol, XDTMQSROBMDMFD-UHFFFAOYSA-N, cyclohexane, 6.5 deg.C, 3814, 84.162, Amber glass bottle, Liquid, 8078, C1CCCCC1, 2.9, 104mbar at 20 deg.C, 0.94 mPaS at 20 deg.C

Specifications

Specifications

6.5°C
Colorless
25 L
Liquid
104 mbar at 20°C
0.94 mPaS at 20°C
110-82-7
81°C
Metal Can
C6H12
3814
84.15g/mol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Cyclohexane

Signal Word
  • Danger

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Flammable liquid Category 2
  • Aspiration toxicity Category 1
  • Skin corrosion/irritation Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H410-Very toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

SDS Documents

Documents

Certificates
Special Offers

Special Offers