missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclohexane, BAKER, J.T. Baker™
86.45 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
1 L
2.5 L
Description
A grade of chemicals for routine research usually controlled by physical criteria and often by assay (by GC).
Specifications
Melting Point | 6.4°C |
Color | Colorless |
Linear Formula | CH2(CH2)4CH2 |
Solubility Information | 0.05g/L in water |
Formula Weight | 84.16g/mol |
Percent Purity | ≥99% |
Grade | BAKER™ |
Density | 0.78g/cm3 at 20°C |
Boiling Point | 81°C |
Quantity | 1 L |
Show More |
Safety and Handling
Signal Word
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Aspiration toxicity Category 1
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P331-Do NOT induce vomiting.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P370+P378-In case of fire: Use to extinguish.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'