missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclohexane, 99+%, for spectroscopy ACS, Thermo Scientific Chemicals
33.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
30.86 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "22914" to get your promotional price.
30.86 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For UV/VIS spectroscopy
Specifications
Melting Point | 6.5°C |
Absorbance | 0.01 max. at 300nm, 0.01 max. at 400nm, 0.02 max. at 260nm, 0.03 max. at 250nm, 0.08 max. at 240nm, 0.2 max. at 230nm, 0.5 max. at 220nm, 1 max. at 210nm |
Density | 0.7700g/mL |
Boiling Point | 81.0°C |
Quantity | 500 mL |
Flash Point | −18°C |
Packaging | Glass Bottle |
Physical Form | Liquid |
Refractive Index | 1.4250 to 1.4270 |
Assay Percent Range | 99+% |
Show More |
Safety and Handling
Product Identifier
- Cyclohexane
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 1
- Flammable liquid Category 2
- Aspiration toxicity Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H410-Very toxic to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'