missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexane, 99.8%, for HPLC, Thermo Scientific Chemicals

Catalog Number p-217364
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
110-82-7
C6H12
84.16
MFCD00003814
XDTMQSROBMDMFD-UHFFFAOYSA-N
hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane
8078
CHEBI:29005
cyclohexane
C1CCCCC1
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
110-82-7
C6H12
84.16
MFCD00003814
XDTMQSROBMDMFD-UHFFFAOYSA-N
hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane
8078
CHEBI:29005
cyclohexane
C1CCCCC1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For HPLC
TRUSTED_SUSTAINABILITY
Melting Point 6.5°C
Absorbance (1 cm cell vs water), (1 cm cell vs water) at 0nm, 0.004 max. at 240nm, 0.004 max. at 250nm, 0.018 max. at 230nm, 0.097 max. at 220nm, 0.201 max. at 210nm, 0.301 max. at 200nm
Color White
Density 0.7700g/mL
Boiling Point 81.0°C
Quantity 1 L
Flash Point −18°C
Packaging Glass Bottle
Physical Form Crystalline Solid
Refractive Index 1.4250 to 1.4270
Assay Percent Range 99.8%
Residue after Evaporation 0.0002 % max.
Specific Gravity 0.77
Linear Formula C6H12
Vapor Pressure Vapor Pressure: 104mbar at 20°C
Viscosity 0.94 mPa.s (20°C)
Beilstein 05, 20
Water 0.01% Max.
Merck Index 15, 2716
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in methanol (57 g/100 mL at 20°C), ethanol, , diethyl ether, acetone, benzene, ccl4.
Formula Weight 84.15
Percent Purity 99.8%
Grade HPLC
Chemical Name or Material Cyclohexane
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cyclohexane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Aspiration toxicity Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'