missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ (Cyclohexadiene)iron tricarbonyl, 97+%

Brand:  Alfa Aesar™ 039403.02

Code : NEW

Additional Details : CAS Number : 12152-72-6 Weight : 0.00050kg Transport : UN number : 3281 Chem class : 6.1 Pack group : II


Quantity 0.5g

 View more versions of this product

Product Code. 11328188

  • £61.90 / 0.50g


Estimated Shipment date
from Supplier 21-06-2018
Add to basket


Specifications

Chemical Identifiers

(Cyclohexadiene)iron tricarbonyl
220.01
UN3281
12152-72-6
MFCD00070435
Tricarbonylcyclohexadiene iron

Specifications

(Cyclohexadiene)iron tricarbonyl
8°C
12152-72-6
≥97%
MFCD00070435
Tricarbonylcyclohexadiene iron
Air Sensitive
0.5g
Liquid
220.01
UN3281
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Tricarbonylcyclohexadiene iron

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 2

Hazard Statement
  • H301-Toxic if swallowed.
  • H330-Fatal if inhaled.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P320-Specific treatment is urgent (see on this label).
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

SDS