Learn More
Cyclofenil, Tocris Bioscience™
Selective estrogen receptor modulator (SERM)
Brand:  Tocris (Bio-Techne) 399950
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13433954
  • £320.46 / 50mg
Estimated Shipment date
from Supplier 24-02-2020
Description
Description
Non-steroidal selective estrogen receptor modulator (SERM). Displays higher affinity for estrogen receptor β (ERβ) than for ERα.
Specifications
Chemical Identifiers
2624-43-3
364.441
cyclofenil, ondogyne, cyclofenyl, cyclopenil, cyclophenyl, fertodur, cyclophenil, sanocrisin, ondonid, rehibin
[4-[(4-acetyloxyphenyl)-cyclohexylidenemethyl]phenyl] acetate
C23H24O4
GVOUFPWUYJWQSK-UHFFFAOYSA-N
2898
CC(=O)OC1=CC=C(C=C1)C(=C2CCCCC2)C3=CC=C(C=C3)OC(=O)C
Specifications
50mg
2624-43-3
cyclofenil, ondogyne, cyclofenyl, cyclopenil, cyclophenyl, fertodur, cyclophenil, sanocrisin, ondonid, rehibin
CC(=O)OC1=CC=C(C=C1)C(=C2CCCCC2)C3=CC=C(C=C3)OC(=O)C
364.441
364.43
Cyclofenil
C23H24O4
GVOUFPWUYJWQSK-UHFFFAOYSA-N
[4-[(4-acetyloxyphenyl)-cyclohexylidenemethyl]phenyl] acetate
2898
>99%