missing translation for 'onlineSavingsMsg'
Learn More

α-Cyclodextrin hydrate, 98+%, Thermo Scientific Chemicals

Catalog Number p-3736063
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
51211-51-9
C36H60O30
972.85
MFCD00150804
HFHDHCJBZVLPGP-UHFFFAOYNA-N
alpha-cyclodextrin hydrate, water.beta-cyclodextrin, cyclohexaamylose hydrate, ^a-cyclodextrin hydrate, alpha-cyclodextrin water, 972.86 anhydrous
60161466
OCC1OC2OC3C(CO)OC(OC4C(CO)OC(OC5C(CO)OC(OC6C(CO)OC(OC7C(CO)OC(OC1C(O)C2O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O
This item is not returnable. View return policy
51211-51-9
C36H60O30
972.85
MFCD00150804
HFHDHCJBZVLPGP-UHFFFAOYNA-N
alpha-cyclodextrin hydrate, water.beta-cyclodextrin, cyclohexaamylose hydrate, ^a-cyclodextrin hydrate, alpha-cyclodextrin water, 972.86 anhydrous
60161466
OCC1OC2OC3C(CO)OC(OC4C(CO)OC(OC5C(CO)OC(OC6C(CO)OC(OC7C(CO)OC(OC1C(O)C2O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chelation/Complexation reagent
TRUSTED_SUSTAINABILITY
CAS Min % 98.0
CAS Max % 100.0
Melting Point 278°C
Color White
Loss on Drying 11% max.
Infrared Spectrum Authentic
Assay Percent Range 0.5% max. Linear oligomers (HPLC)
Packaging Glass bottle
Quantity 5 g
Merck Index 14, 2718
Specific Rotation + 148
Solubility Information Solubility in water: 145g/L (25°C)
IUPAC Name 5,10,15,20,25,30-hexakis(hydroxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29-dodecaoxaheptacyclo[26.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶]dotetracontane-31,32,33,34,35,36,37,38,39,40,41,42-dodecol
Formula Weight 972.85
Percent Purity 98+%
Physical Form Crystalline Powder
Chemical Name or Material α-Cyclodextrin hydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'