missing translation for 'onlineSavingsMsg'
Learn More

Cyclobutanecarboxylic acid, 98%, Thermo Scientific Chemicals

Catalog Number 10468250
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
3721-95-7
C5H8O2
100.12
MFCD00001323
TXWOGHSRPAYOML-UHFFFAOYSA-N
cyclobutylcarboxylic acid, cyclo-butyl formic acid, cyclobutane carboxylic acid, unii-0rdj7c51o0, pubchem14139, acmc-1cnqb, cyclobutan carboxylic acid, cyclobutyl carboxylic acid, cyclobutancecarboxylic acid, cyclobutane-carboxylic acid
19494
cyclobutanecarboxylic acid
OC(=O)C1CCC1
This item is not returnable. View return policy
3721-95-7
C5H8O2
100.12
MFCD00001323
TXWOGHSRPAYOML-UHFFFAOYSA-N
cyclobutylcarboxylic acid, cyclo-butyl formic acid, cyclobutane carboxylic acid, unii-0rdj7c51o0, pubchem14139, acmc-1cnqb, cyclobutan carboxylic acid, cyclobutyl carboxylic acid, cyclobutancecarboxylic acid, cyclobutane-carboxylic acid
19494
cyclobutanecarboxylic acid
OC(=O)C1CCC1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -20.0°C
Color Colorless to Yellow
Density 1.0400g/mL
Boiling Point 195.0°C
Flash Point 83°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C4H7CO2H
Refractive Index 1.4410 to 1.4450
Quantity 5 g
Beilstein 09, 5
Specific Gravity 1.04
Formula Weight 100.12
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Cyclobutanecarboxylic acid
Show More Show Less
compliance-icons
Product Identifier
  • Cyclobutanecarboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'