missing translation for 'onlineSavingsMsg'
Learn More

Cyanuric chloride, 99%, Thermo Scientific Chemicals

Catalog Number p-95384
19.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic drum
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
108-77-0
C3Cl3N3
184.40
MFCD00006046
MGNCLNQXLYJVJD-UHFFFAOYSA-N
cyanuric chloride, cyanuryl chloride, cyanuric acid chloride, s-triazine trichloride, trichlorocyanidine, cyanuric trichloride, tricyanogen chloride, sym-trichlorotriazine, cyanur chloride, trichloro-s-triazine
7954
CHEBI:58964
ClC1=NC(Cl)=NC(Cl)=N1
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
108-77-0
C3Cl3N3
184.40
MFCD00006046
MGNCLNQXLYJVJD-UHFFFAOYSA-N
cyanuric chloride, cyanuryl chloride, cyanuric acid chloride, s-triazine trichloride, trichlorocyanidine, cyanuric trichloride, tricyanogen chloride, sym-trichlorotriazine, cyanur chloride, trichloro-s-triazine
7954
CHEBI:58964
ClC1=NC(Cl)=NC(Cl)=N1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY
Melting Point 145.0°C to 148.0°C
Color Beige-Brown to Yellow
Density 1.9200g/mL
Boiling Point 190.0°C
Flash Point >200°C
Infrared Spectrum Authentic
Assay Percent Range 0.99
Packaging Glass bottle
Quantity 250 g
Beilstein 26,35
Fieser 03,72; 04,522; 05,687; 06,149; 10,114; 15,105
Merck Index 152691
Specific Gravity 1.92
Solubility Information Solubility in water: reacts. Other solubilities: reacts with ethanol, 250g/L acetone (25°C), soluble in chloroform, hot ether, dioxane, ketones, and carbontetrachloride
IUPAC Name trichloro-1,3,5-triazine
Viscosity 0.57 mPa.s (150°C)
Formula Weight 184.41
Percent Purity 0.99
Physical Form Solidified Mass or Fragments
Chemical Name or Material Cyanuric chloride, 0.99
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Cyanuric chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
  • Acute toxicity Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H330-Fatal if inhaled.
  • H335-May cause respiratory irritation.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'