missing translation for 'onlineSavingsMsg'
Learn More

Cyanopropylmethyldichlorosilane, 97%, Thermo Scientific™

Catalog Number p-4229155
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
1190-16-5
C5H9Cl2NSi
182.12
MFCD00019889
UIFBMBZYGZSWQE-UHFFFAOYSA-N
3-cyanopropyl methyldichlorosilane, 3-cyanopropylmethyldichlorosilane, 4-dichloromethylsilyl-butyronitrile, 3-cyanopropyldichloromethylsilane, butanenitrile, 4-dichloromethylsilyl, unii-a09zd6eimj, 4-dichloromethylsilyl butyronitrile, methylcyanopropyldichlorosilane, 4-dichloro methyl silyl butanenitrile, dichlor-3-kyanpropyl-methylsilan
14479
4-[dichloro(methyl)silyl]butanenitrile
C[Si](Cl)(Cl)CCCC#N
This item is not returnable. View return policy
1190-16-5
C5H9Cl2NSi
182.12
MFCD00019889
UIFBMBZYGZSWQE-UHFFFAOYSA-N
3-cyanopropyl methyldichlorosilane, 3-cyanopropylmethyldichlorosilane, 4-dichloromethylsilyl-butyronitrile, 3-cyanopropyldichloromethylsilane, butanenitrile, 4-dichloromethylsilyl, unii-a09zd6eimj, 4-dichloromethylsilyl butyronitrile, methylcyanopropyldichlorosilane, 4-dichloro methyl silyl butanenitrile, dichlor-3-kyanpropyl-methylsilan
14479
4-[dichloro(methyl)silyl]butanenitrile
C[Si](Cl)(Cl)CCCC#N
Color Colorless to Yellow
Density 1.1500g/mL
Boiling Point 93.0°C to 94.0°C (8.0 mmHg)
Flash Point 92°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4570 to 1.4590 (20°C, 589nm)
Quantity 5 mL
Specific Gravity 1.15
Formula Weight 182.13
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Cyanopropylmethyldichlorosilane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Cyanopropylmethyldichlorosilane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
  • EUH014-Reacts violently with water.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'