missing translation for 'onlineSavingsMsg'
Learn More

Cyanogen bromide, 97%, ACROS Organics™

Brand:  Acros Organics 110785000

Code : 29

Additional Details : CAS Number : 506-68-3 Weight : 0.60000kg Transport : UN number : 1889 Chem class : 1 Pack group : I


Quantity 500g

 View more versions of this product

Product Code. 10465852

  • £199.10 / 500g


EU Stock 28
Add to basket

Sea freight only: For EU stock allow up to 15 days delivery.

Specifications

Chemical Identifiers

Cyanogen bromide
506-68-3
97%
BrCN
MFCD00011597
97%
97%
CBrN
105.93
Bromine cyanide

Specifications

Cyanogen bromide
>65°C
White
500g
506-68-3
97%
CBrN
105.93
01,174; 02,93; 04,110; 05,169; 06,148; 08,131; 09,127
Bromine cyanide
1.44
61.0° to 62.0°C
49.0° to 54.0°C
97%
Crystals
97%
BrCN
MFCD00011597
15, 2683
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Cyanogen bromide

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H310-Fatal in contact with skin.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H314-Causes severe skin burns and eye damage.
  • H330-Fatal if inhaled.
  • H300-Fatal if swallowed.

Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS