missing translation for 'onlineSavingsMsg'
Learn More

Cyanogen bromide, 97%, ACROS Organics™

Brand:  Acros Organics 110780250

Code : 29

Additional Details : CAS Number : 506-68-3 Weight : 0.20304kg Transport : UN number : 1889 Chem class : 6.1 Pack group : I


Packaging Glass Bottle in Tin Can
Quantity 25g

 View more versions of this product

Product Code. 10040770

  • £21.90 / 25g

Estimated Shipment
28-04-2020

Please to purchase this item. Need a web account? Register with us today!


Sea freight only: For EU stock allow up to 15 days delivery.

Description

Description

C-X bond formation reagent
Specifications

Chemical Identifiers

506-68-3
105.922
ATDGTVJJHBUTRL-UHFFFAOYSA-N
10476
C(#N)Br
CBrN
MFCD00011597
cyanogen bromide, bromine cyanide, bromocyanide, cyanobromide, bromocyanogen, bromocyan, campilit, cyanogen monobromide, bromure de cyanogen, cyanogen bromide brcn
carbononitridic bromide

Specifications

Cyanogen bromide
Glass Bottle in Tin Can
25g
506-68-3
CBrN
MFCD00011597
15, 2683
ATDGTVJJHBUTRL-UHFFFAOYSA-N
carbononitridic bromide
10476
Crystals
1.44
White
97%
97%
BrCN
01,174; 02,93; 04,110; 05,169; 06,148; 08,131; 09,127
cyanogen bromide, bromine cyanide, bromocyanide, cyanobromide, bromocyanogen, bromocyan, campilit, cyanogen monobromide, bromure de cyanogen, cyanogen bromide brcn
C(#N)Br
105.922
105.93
≥96.0%
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Cyanogen bromide

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H310-Fatal in contact with skin.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H314-Causes severe skin burns and eye damage.
  • H330-Fatal if inhaled.
  • H300-Fatal if swallowed.

Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS Documents