Learn More
Cyanogen bromide, 3M solution in dichloromethane, AcroSeal™, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 428911000
Code : 29
Additional Details : CAS Number : 506-68-3 Weight : 0.14400kg Transport : UN number : 2927 Chem class : 6.1 Pack group : I
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Cyanogen bromide | |
506-68-3,75-09-2 | |
100 mL | |
MFCD00011597 | |
cyanogen bromide, bromine cyanide, bromocyanide, cyanobromide, bromocyanogen, bromocyan, campilit, cyanogen monobromide, bromure de cyanogen, cyanogen bromide brcn | |
C(#N)Br | |
105.93 | |
105.93 | |
AcroSeal glass Bottle packed in tin can | |
1.4400g/mL |
Colorless to Yellow | |
Liquid | |
BrCN | |
15, 2683 | |
ATDGTVJJHBUTRL-UHFFFAOYSA-N | |
carbononitridic bromide | |
10476 | |
2.95 to 3.30M | |
1.44 |
Safety and Handling
- Cyanogen bromide, 3M solution in dichloromethane
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 1
- Acute toxicity Category 2
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
Hazard Statement
- H310-Fatal in contact with skin.
- H318-Causes serious eye damage.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H314-Causes severe skin burns and eye damage.
- H330-Fatal if inhaled.
- H351-Suspected of causing cancer.
- H336-May cause drowsiness or dizziness.
- H300-Fatal if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only