Learn More
CY 208-243, Tocris Bioscience™
Selective D1-like agonist
Brand:  Tocris (Bio-Techne) 124950
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13488973
  • £646.37 / 50mg
Estimated Shipment
03-10-2018
Add to basket
Description
Description
Centrally active dopamine D1 receptor agonist, selective over D2 receptor sites. Stimulates adenylate cyclase in rat striatal homogenates with an EC50 of 125 nM. Unlike SKF 38393 (Cat. No. 0922), it exerts antiParkinsonian activity in animal models.
Specifications
Chemical Identifiers
100999-26-6
274.367
(-)-(6aR,12bR)-4,6,6a,7,8,12b-Hexahydro-7-methylindolo[4,3-a]phenanthridin
C19H18N2
WRNKIDLXXXIELU-IEBWSBKVSA-N
58144
Specifications
CY 208-243
100999-26-6
C19H18N2
WRNKIDLXXXIELU-IEBWSBKVSA-N
274.367
274.36
50mg
>98%
(-)-(6aR,12bR)-4,6,6a,7,8,12b-Hexahydro-7-methylindolo[4,3-a]phenanthridin
CN1CC2=CC=CC=C2C3C1CC4=CNC5=CC=CC3=C45
58144
Videos