missing translation for 'onlineSavingsMsg'
Learn More

CXCL9 (MIG), PE, clone: MIG-2F5.5, eBioscience™

Armenian Hamster Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-3009-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15228739

  • £80.50 / 25µg


EU Stock 11
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The monoclonal antibody MIG-2F5.5 reacts with mouse monokine induced by IFN gamma (mig, mig-1, CXCL9). This 14.4kDa inflammatory chemokine is specifically induced by IFN gamma, but not other activators such as LPS or IFN alpha. Secretion of mig, mainly by macrophages results in the chemotaxis of a variety of activated T cells via the CXCR3 chemokine receptor. Mig is involved many areas of research including autoimmune diseases, cancer, and inflammation.Applications Reported: This MIG-2F5.5 antibody has been reported for use in intracellular staining followed by flow cytometric analysis.Applications Tested: This MIG-2F5.5 antibody has been tested by intracellular staining followed by flow cytometric analysis of IFN gamma-stimulated thioglycolate-elicited peritoneal exudate cells and RAW 264.7 cells. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CXCL9 (MIG)
MIG-2F5.5
PE
Conjugated
Armenian Hamster
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry (Intracellular Staining)
0.2mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
monokine induced by IFN-g
IgG
RUO
Primary
Murine
Videos