missing translation for 'onlineSavingsMsg'
Learn More

CX3CR1, PerCP-eFluor™ 710, clone: 2A9-1, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 46-6099-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15529336

  • £97.68 / 25 tests


Estimated Shipment date
from Supplier 30-05-2018
Add to basket

Description

Description

Description: This 2A9-1 monoclonal antibody reacts with human CX3CR1, which is the receptor for fractalkine, a transmembrane chemokine of the CX3C family. CX3CR1 is a seven transmembrane G protein-coupled receptor expressed on CD16+ NK cells, T cells (e.g. CD8+, CD4+, and gamma/delta), and monocytes. In non-immune cells, CX3CR1 has been found on osteoclast precursors and microglia. Little to no CX3CR1 surface expression can be detected on B cells and granulocytes. Together, fractalkine and its receptor mediate cell-cell adhesion and chemotaxis of NK cells, T cells, and monocytes. The expression of CX3CR1 has also been correlated with high levels of intracellular perforin and granzyme B.Applications Reported: This 2A9-1 antibody has been reported for use in flow cytometric analysis.Applications Tested: This 2A9-1 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. PerCP-eFluor® 710 can be used in place of PE-Cy5, PE-Cy5.5 or PerCP-Cy5.5. PerCP-eFluor® 710 emits at 710 nm and is excited with the blue laser (488 nm). Please make sure that your instrument is capable of detecting this fluorochrome. For a filter configuration, we recommend using the 685 LP dichroic mirror and 710/40 band pass filter, however the 695/40 band pass filter is an acceptable alternative. Our testing indicates that PerCP-eFluor® 710 conjugated antibodies are stable when stained samples are exposed to freshly prepared 2% formaldehyde overnight at 4°C, but please evaluate for alternative fixation protocols.Excitation: 488 nm; Emission: 710 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CX3CR1
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P49238
CCRL1, CMKBRL1, CMKDR1, GPR13, GPRV28, V28
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
2A9-1
PerCP-eFluor 710
CX3CR1
Conjugated
CX3CR1
IgG2b, kappa
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
1524
Videos

For Research Use Only.