missing translation for 'onlineSavingsMsg'
Learn More

CX3CR1, APC, clone: 2A9-1, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 17-6099-42

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15538176

  • £186.79 / 100 tests


EU Stock 4
Add to basket

Description

Description

Description: This 2A9-1 monoclonal antibody reacts with human CX3CR1, which is the receptor for fractalkine, a transmembrane chemokine of the CX3C family. CX3CR1 is a seven transmembrane G protein-coupled receptor expressed on CD16+ NK cells, T cells (e.g. CD8+, CD4+, and gamma/delta), and monocytes. In non-immune cells, CX3CR1 has been found on osteoclast precursors and microglia. Little to no CX3CR1 surface expression can be detected on B cells and granulocytes. Together, fractalkine and its receptor mediate cell-cell adhesion and chemotaxis of NK cells, T cells, and monocytes. The expression of CX3CR1 has also been correlated with high levels of intracellular perforin and granzyme B.Applications Reported: This 2A9-1 antibody has been reported for use in flow cytometric analysis.Applications Tested: This 2A9-1 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CX3CR1
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P49238
CCRL1, CMKBRL1, CMKDR1, GPR13, GPRV28, V28
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
2A9-1
APC
CX3CR1
Conjugated
CX3CR1
IgG2b, kappa
100 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
1524
Videos

For Research Use Only.