missing translation for 'onlineSavingsMsg'
Learn More

Duran™ Culture Tubes With ISO Thread

Brand:  Duran™ 261352151

Code : NEW

Additional Details : Weight : 1.70000kg

 View more versions of this product

Product Code. 15232079

  • £197.00 / Pack of 50


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications

Specifications

16mm
ISO
Screw
160mm
50
50Pack
Yes
Screw
160 L x 16 dia.mm
Cap
18
Culture Tube
Videos