missing translation for 'onlineSavingsMsg'
Learn More

Duran™ Culture Tubes

Brand:  Duran™ 261352402

Code : A0

Additional Details : Weight : 1.35000kg

 View more versions of this product

Product Code. 11650039

  • £140.00 / Pack of 50


Estimated Shipment
14-02-2018
Add to basket

Specifications

Specifications

20mL
20 x 150mm
50
50Pack
20mm
150mm
18
Culture Tube
Videos