Learn More
Sartorius™ Cubis™ Series MSA1202S Model Precision Balances
Top-of-the-line technology and information design
£5130.00 - £5371.00
Specifications
Connection Type RS-232 25-pin / Bluetooth / RS-232 9-pin, incl. PS
Corner Load 20 mg
Display Type High-res Color TFT for High-contrast Display
Readability 10 mg
Format Kilograms
View More Specs
Products 2
Product Code Brand Readability Model Stabilization Time Price Quantity & Availability  
Product Code Brand Readability Model Stabilization Time Price Quantity & Availability  
10278004
View accessory products View Documents Promotion Details
Sartorius™
MSA1202S-000-D0
10 mg MSA1202S 0.8 sec. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11858451
View accessory products View Documents Promotion Details
Sartorius™
MSA1202S-0CE-D0
10 mg MSA1202S 0.8 sec. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description
Description
The MSA display and control unit is the top of the line for operation of the Cubis. The high-resolution graphic display and touch screen provide an optimal display and control unit for the most stringent requirements. Cubis MSA models are equipped with an intelligent warning function as a standard feature which detects the incline immediately. Clear adjustment prompts on the display support the user when levelling the balance. Perfect communication is ensured by three data interfaces included as standard: Ethernet with implemented Web services, USB PC interface and a RS232C interface for Sartorius accessories/printers. Optional interfaces include Bluetooth, 9-pin RS232C PC and PS/2 interfaces. Electrical supply 100V to 240V, 50/60Hz, a. c. adapter. The built-in SD card reader can be used to transfer weighing data, user profiles, task configurations and settings from one balance to another or to a PC. Measures weight, density determination, averaging, formulation, % weighing, check weighing and totalising.
  • Tiltable and removable display and control unit
  • Compensates for eccentric/off centre loading error
  • Motorised draft shield
  • Password protected
  • Audit trail function
  • GLP compliant
  • Optional motorised levelling function
  • Mass unit conversion
  • Time controlled functions
  • Second tare memory
Specifications
Specifications
RS-232 25-pin / Bluetooth / RS-232 9-pin, incl. PS
High-res Color TFT for High-contrast Display
Kilograms
Assisted
≤1 sec.
MSA1202S
≤±7 mg
5°C to 40°C
Precision Balance
193 mm
172 mm
191 mm
100/240 V 50/60 Hz 1 A 7 w 15 VDC ±5%
7 w
50/60 Hz
20 mg
10 mg
50 to 80% RH
≤±20 mg
10 g
–1200 g
0.8 sec.
500 g
191 x 193 x 172 mm
206 mm
172 mm
206 mm
100/240 V
1 A