missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Key Industrial Equipment™ Cube locker, grey/light grey, Ekwo, 380mm x 380mm x 380mm
Brand: Key Industrial Equipment™ A327782
Code : Z3
Additional Details : Weight : 6.00000kg
Description
Strong steel lockers ideal for stacking, fixing side-by-side or back-to-back. Strong, lockable, hinged doors fitted with label holder and nameplate for personal identification, doors open to 180° for easy access. Cylinder lock supplied with two keys.- Light grey door
- Dimensions (H x W x D) : 380mm x 380mm x 380mm