missing translation for 'onlineSavingsMsg'
Learn More

Sino Biological™ CSF3R (CD114) Recombinant Human Protein, His Tag

Expressed from DNA sequence encoding extracellular domain (Met 1-Pro 621) of human CSF3R precursor fused topolyhistidine tag at C-terminus

Brand:  Sino Biological™ 10218H08H50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13415826

  • £255.05 / 50µg


Estimated Shipment
12-02-2018
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

  • Measured by its ability to inhibit the CSF3 induced proliferation of NFS60 mouse myeloid cells. The ED50 for this effect is typically 20-80 ng/ml.
Specifications

Specifications

Studying protein-protein interactions and protein structure analysis
1441
>85% by SDS-PAGE
Human
Truncated, Cell Surface Protein, Recombinant
Colony stimulating factor 3 receptor
Assay Standard
Mammalian
His Tag
Lyophilized
68kd
50μg
−20° to −70°C
<1 EU/μg
CSF3R
Cytokines and Receptors
CSF3R (CD114) Recombinant Human Protein
Videos