missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Crystal Violet lactone

Brand:  Alfa Aesar™ 043710.14

Code : NEW

Additional Details : CAS Number : 1552-42-7 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 11300318

  • £126.30 / 25g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

Crystal Violet lactone
C26H29N3O2
MFCD00070611
1552-42-7
415.53

Specifications

Crystal Violet lactone
Powder
415.53
180° to 183°C
1552-42-7
C26H29N3O2
MFCD00070611
25g
Videos
SDS