missing translation for 'onlineSavingsMsg'
Learn More

Saint Gobain Performance Plastics™ Crystal Clear Tygon™ S3™ E-LFL Tubing With 2-stop Configuration

Brand:  Saint Gobain Performance Plastics™ 070602-07-ND

Code : 02

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 15181723

  • £74.29 / Pack of 12


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications

Specifications

USP Class VI, ISO 10993 and FDA
Crystal Clear
For use in peristaltic pump applications
56ShA
–51°C
0.76mm
Tygon S3 E-LFL
0.85mm
Black/Black
Flexible
For peristaltic pump
74°C
12 Pack
2.46mm
381mm
Videos