missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Crystal Clear (TPX™) PMP Volumetric Flasks with Stopper

Brand:  Kartell™ 2486

Code : 02

Additional Details : Weight : 1.50000kg

 View more versions of this product

Product Code. 11398533

Quantity Price
1 £8.10 / Each
10 £7.41 / Each
In Stock 2

Add to basket


Specifications

Specifications

Yes
Crystal clear
100mL
174mm
Stopper
58mm
TPX™
Stopper
Videos