missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Crystal Clear (TPX™) PMP Volumetric Flasks with Stopper

Brand:  Kartell™ 2485

Code : 02

Additional Details : Weight : 0.01316kg

 View more versions of this product

Product Code. 11388533

  • £7.15 / Each


In Stock 18
Add to basket

Specifications

Specifications

Yes
Crystal clear
50mL
143mm
Stopper
46mm
TPX™þPMP (Polymethylpentene)
Stopper
Videos