missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Crystal Clear (TPX™) PMP Volumetric Flasks with Screw Cap

Brand:  Kartell™ 2491

Code : 03

Additional Details : Weight : 0.01692kg

Product Code. 11301185

  • £12.44 / Each


In Stock 6
Add to basket

You may only order up to the available stock. For more details call customer services on 01509 555 500

Description

Description

These flasks create no meniscus so final readings are easy. Thick walls eliminate distortion and thin neck make final reading precise. Chemically non-absorbent. Autoclavable.
Specifications

Specifications

Yes
Crystal clear
100mL
174mm
20Pack
Screw cap
58mm
TPX™
Screw cap
Videos
Documentation