missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Cryptic Antibody, R&D Systems™
Mouse Monoclonal Antibody
POA
Specifications
Antigen | CYRPTIC/CFC1 |
---|---|
Clone | 219708 |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Classification | Monoclonal |
Description
CYRPTIC/CFC1 Monoclonal specifically detects CYRPTIC/CFC1 in Human samples. It is validated for Western Blot.Specifications
CYRPTIC/CFC1 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CFC1, CFC-1, CFC1B, cripto, FRL-1, cryptic family 1, Cryptic family protein 1, cryptic protein, CRYPTICFLJ77897, HTX2MGC133213 | |
CFC1 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
219708 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
P0CG36 | |
55997 | |
Mouse myeloma cell line NS0-derived recombinant human Cryptic Tyr26-Gly169 Accession # P0CG36 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Cryptic in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse Cryptic is observed. |
For Research Use Only