Learn More
SPEX Certiprep™ Cryogenic Gloves
Click to view available options
Size:
Small
Medium
Large
Description
- Cryogenic gloves specifically designed to protect hands and arms from the hazards associated with liquid nitrogen handling
- Highly recommended for the safe operation of the Freezer/Mill™
- Multi-layer insulation for optimal protection
- Flexible and lightweight to allow excellent dexterity
- Comfortable to wear for extended periods of time
Specifications
Color | Blue |
Material | Fabric |
Powdered | Powder Free |
Product Type | Cryogenic Gloves |
Size | Large |
Sterility | Non-sterile |