missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tempshield™ Cryo-Gloves™ Blue Shoulder-Length Cryogenic Gloves
State-of-the art materials and multi-layered construction provide cryogenic protection for ultra-cold applications down to -196°C (-320°F).
£464.00
Specifications
Certifications/Compliance (Europe) | EN 420, EN 511 |
---|---|
Description | Shoulder Length |
Color | Blue |
Hand | Specific |
Product Type | Cryogenic Gloves |
Alert:
Not for immersion in liquid nitrogen, for use near open flame, or in hazardous areas where ignition could occur.
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
11766419
|
Tempshield™
SHL |
Large |
£464.00
1 pair |
Estimated Shipment: 01-08-2024
Log in to see stock. |
|||||
16072252
|
Tempshield™
SHXL |
X-Large |
£464.00
1 pair |
Estimated Shipment: 01-08-2024
Log in to see stock. |
|||||
Description
State-of-the art materials and multi-layered construction provides cryogenic protection for ultra-cold applications down to -196°C (-320°F).
- Cryogenic protection for ultra-cold applications down to -196°C (-320°F)
- State-of-the art materials and multi-layered construction allow for a maximum level of thermal protection, flexibility, and dexterity
- High performance, thermal lining wicks moisture away from hands to maintain comfort over extended periods of wear
- Ideal for applications where function is important and safety is critical
- Not intended for immersion in liquid nitrogen or other cryogenic liquids
Specifications
EN 420, EN 511 | |
Blue | |
Cryogenic Gloves | |
Dry Ice Handling, Low/Ultra-Low Temperature Freezers, and Closed Cryogenic Systems | |
-320°F | |
24.5 to 27.3 in. |
Shoulder Length | |
Specific | |
Yes | |
Cryo-Gloves™ | |
-196°C | |
62 to 69.5 cm |