Learn More
Crotyl chloride, 70%, tech., Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 154805000
Code : 29
Additional Details : CAS Number : 591-97-9 Weight : 0.51000kg Transport : UN number : 2924 Chem class : 3 Pack group : II
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Crotyl chloride | |
591-97-9,563-52-0,1190-22-3 | |
Colorless | |
80.0°C to 85.0°C | |
Authentic | |
Glass bottle | |
1.4300 to 1.4360 | |
MFCD00000987 | |
01, II, 176 | |
15, 2130 | |
Solubility in water:+/- 14g/L (20°C) | |
0.46 mPa.s (20°C) | |
70% | |
Liquid |
tech., 70% | |
-65.0°C | |
0.9200g/mL | |
−15°C | |
65% min. (GC) | |
C4H7Cl | |
CH3CH=CHCH2Cl | |
500 mL | |
0.92 | |
1-Chloro-2-butene | |
90.55 | |
90.55 | |
Technical |
Safety and Handling
- Crotyl chloride
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only