missing translation for 'onlineSavingsMsg'
Learn More

Crotonic acid, 98%, Thermo Scientific Chemicals

Catalog Number 10615972
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
107-93-7
C4H6O2
86.09
MFCD00002701
LDHQCZJRKDOVOX-NSCUHMNNSA-N
crotonic acid, e-but-2-enoic acid, but-2-enoic acid, trans-2-butenoic acid, trans-crotonic acid, 3-methylacrylic acid, e-2-butenoic acid, 2-butenoic acid, 2e-but-2-enoic acid, e-crotonic acid
637090
CHEBI:41131
(E)-but-2-enoic acid
CC=CC(=O)O
This item is not returnable. View return policy
107-93-7
C4H6O2
86.09
MFCD00002701
LDHQCZJRKDOVOX-NSCUHMNNSA-N
crotonic acid, e-but-2-enoic acid, but-2-enoic acid, trans-2-butenoic acid, trans-crotonic acid, 3-methylacrylic acid, e-2-butenoic acid, 2-butenoic acid, 2e-but-2-enoic acid, e-crotonic acid
637090
CHEBI:41131
(E)-but-2-enoic acid
CC=CC(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 72.0°C
Color White to Yellow
Boiling Point 185.0°C
Flash Point 88°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Plastic bottle
Linear Formula CH3CH=CHCO2H
Quantity 500 g
Beilstein 02, IV, 1491
Merck Index 15, 2588
Solubility Information Solubility in water: 94g/L (25°C). Other solubilities: soluble in ethanol,acetone and toluene
Formula Weight 86.09
Percent Purity 98%
Physical Form Crystalline Flakes
Chemical Name or Material Crotonic acid, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Crotonic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'