missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Crossveinless-2/CV-2 Antibody, R&D Systems™
Rat Monoclonal Antibody has been used in 1 publication
POA
Specifications
Antigen | Crossveinless-2/CV-2/BMPER |
---|---|
Clone | 349920 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
Crossveinless-2/CV-2/BMPER Monoclonal specifically detects Crossveinless-2/CV-2/BMPER in Mouse samples. It is validated for Western Blot.Specifications
Crossveinless-2/CV-2/BMPER | |
LYOPH | |
Western Blot | |
Unconjugated | |
Rat | |
Mouse | |
AAH66153 | |
168667 | |
Mouse myeloma cell line NS0-derived recombinant mouse Crossveinless-2/CV-2 Val34-Arg685 Accession # AAH66153 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse Crossveinless-2/CV-2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, 100% cross-reactivity with recombinant human CV-2 is observed. |
349920 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
BMP binding endothelial regulator, BMP-binding endothelial regulator precursor protein, BMP-binding endothelial regulator protein, BMPER, Bone morphogenetic protein-binding endothelial cell precursor-derived regulator, CRIM3, crossveinless 2, crossveinless-2, Cv2, CV-2, hCV2, KIAA1965, Protein crossveinless-2 | |
BMPER | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only