missing translation for 'onlineSavingsMsg'
Learn More

Crizotinib, Tocris Bioscience™

Potent c-MET/ALK inhibitor

Brand:  RD Systems (Bio-Techne) 4368/10

Code : Z1

Additional Details : CAS Number : 877399-52-5 Weight : 0.00010kg

Quantity 10mg

 View more versions of this product

Product Code. 13464634

  • £267.88 / 10mg

Estimated Shipment
12-08-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent inhibitor of c-MET and anaplastic lymphoma kinase (ALK) (cell IC50 values are 8.0 and 20 nM respectively). Selective for c-MET and ALK against >120 different kinases. Displays antitumor efficacy in multiple tumor models; inhibits c-MET-dependent proliferation, migration and invasion of human tumor cells in vitro. Orally bioavailable.
Specifications

Chemical Identifiers

877399-52-5
450.339
crizotinib, xalkori, r-crizotinib, crizotinibum, 3-1r-1-2,6-dichloro-3-fluorophenyl ethoxy-5-1-4-piperidinyl-1h-pyrazol-4-yl-2-pyridinamine, xalkori tn, r-3-1-2,6-dichloro-3-fluorophenyl ethoxy-5-1-piperidin-4-yl-1h-pyrazol-4-yl pyridin-2-amine, crizotinib pf-02341066, 3-1r-1-2,6-dichloro-3-fluorophenyl ethoxy-5-1-piperidin-4-yl-1h-pyrazol-4-yl pyridin-2-amine, crizotinib pf-2341066
CHEBI:64310
CC(C1=C(C=CC(=C1Cl)F)Cl)OC2=C(N=CC(=C2)C3=CN(N=C3)C4CCNCC4)N
C21H22Cl2FN5O
KTEIFNKAUNYNJU-GFCCVEGCSA-N
11626560
3-[(1R)-1-(2,6-dichloro-3-fluorophenyl)ethoxy]-5-(1-piperidin-4-ylpyrazol-4-yl)pyridin-2-amine

Specifications

Crizotinib
877399-52-5
C21H22Cl2FN5O
KTEIFNKAUNYNJU-GFCCVEGCSA-N
3-[(1R)-1-(2,6-dichloro-3-fluorophenyl)ethoxy]-5-(1-piperidin-4-ylpyrazol-4-yl)pyridin-2-amine
11626560
450.34
10mg
>99%
crizotinib, xalkori, r-crizotinib, crizotinibum, 3-1r-1-2,6-dichloro-3-fluorophenyl ethoxy-5-1-4-piperidinyl-1h-pyrazol-4-yl-2-pyridinamine, xalkori tn, r-3-1-2,6-dichloro-3-fluorophenyl ethoxy-5-1-piperidin-4-yl-1h-pyrazol-4-yl pyridin-2-amine, crizotinib pf-02341066, 3-1r-1-2,6-dichloro-3-fluorophenyl ethoxy-5-1-piperidin-4-yl-1h-pyrazol-4-yl pyridin-2-amine, crizotinib pf-2341066
CC(C1=C(C=CC(=C1Cl)F)Cl)OC2=C(N=CC(=C2)C3=CN(N=C3)C4CCNCC4)N
450.339
CHEBI:64310
Videos
Documents