missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CRIM1 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 1 publication
POA
Specifications
Antigen | CRIM1 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
CRIM1 Polyclonal specifically detects CRIM1 in Human samples. It is validated for Western Blot.Specifications
CRIM1 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Human | |
CRIM-1, cysteine rich transmembrane BMP regulator 1 (chordin-like), cysteine-rich motor neuron 1, cysteine-rich motor neuron 1 protein, Cysteine-rich repeat-containing protein S52, MGC138194, S52 | |
CRIM1 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
Q9NZV1 | |
51232 | |
Mouse myeloma cell line NS0-derived recombinant human CRIM1 Leu35-Ser939 Accession # Q9NZV1 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human CRIM1 in direct ELISAs and Western blots. |
For Research Use Only