missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cresyl Violet acetate, pure, high purity biological stain

Catalog Number p-30216
63.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
10510-54-0
C18H15N3O3
321.34
MFCD00013151,MFCD00013151
XKOCOMKJPWEOHX-UHFFFAOYSA-M
44134641
acetic acid;5-iminobenzo[a]phenoxazin-9-amine
CC([O-])=O.NC1=CC2=[O+]C3=C(N=C2C=C1)C1=CC=CC=C1C(N)=C3
This item is not returnable. View return policy
10510-54-0
C18H15N3O3
321.34
MFCD00013151,MFCD00013151
XKOCOMKJPWEOHX-UHFFFAOYSA-M
44134641
acetic acid;5-iminobenzo[a]phenoxazin-9-amine
CC([O-])=O.NC1=CC2=[O+]C3=C(N=C2C=C1)C1=CC=CC=C1C(N)=C3

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 140.0°C to 143.0°C
Dye Content 65% min.
Solubility Information Solubility in water: soluble in water. Other solubilities: soluble in ethanol, insoluble in aceton
Infrared Spectrum Authentic
Formula Weight 321.34
Loss on Drying 20% max. (50°C) (0.05mm Hg)
Grade Pure
Packaging Glass bottle
Color Black to Green
Quantity 5 g
Physical Form Fine Crystalline Powder
Chemical Name or Material Cresyl violet acetate
Show More Show Less
compliance-icons
Product Identifier
  • Cresyl Violet acetate
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'