missing translation for 'onlineSavingsMsg'
Learn More

Cresyl Violet acetate, pure, certified, Thermo Scientific Chemicals

Catalog Number 11916941
Click to view available options
Quantity:
2.5 g
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
10510-54-0
C18H15N3O3
321.34
MFCD00013151,MFCD00013151
XKOCOMKJPWEOHX-UHFFFAOYSA-M
44134641
CC([O-])=O.NC1=CC2=[O+]C3=C(N=C2C=C1)C1=CC=CC=C1C(N)=C3
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
10510-54-0
C18H15N3O3
321.34
MFCD00013151,MFCD00013151
XKOCOMKJPWEOHX-UHFFFAOYSA-M
44134641
CC([O-])=O.NC1=CC2=[O+]C3=C(N=C2C=C1)C1=CC=CC=C1C(N)=C3

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 140.0°C to 143.0°C
Absorption Ratio 0.94 to 1.2
Dye Content 65% min.
Solubility Information Solubility in water: soluble in water. Other solubilities: soluble in ethanol, insoluble in aceton
Infrared Spectrum Authentic
IUPAC Name acetic acid;5-iminobenzo[a]phenoxazin-9-amine
Lambda Max 596 to 599nm
Formula Weight 321.34
Grade Pure
Packaging Glass bottle
Color Black to Green
Quantity 50 g
Physical Form Fine Crystalline Powder
Chemical Name or Material Cresyl Violet acetate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'