missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Cresol Red, Water Soluble, Indicator Grade
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
Specifications
Solubility Information | Solubility in water: Soluble |
Infrared Spectrum | Authentic |
Formula Weight | 404.4 |
Grade | Pure |
Packaging | Glass bottle |
Visual Transition Interval | from pH 7.0 (yellow) to pH 8.8 (purple) |
Color | Brown |
Quantity | 5 g |
Physical Form | Powder |
Chemical Name or Material | Cresol Red, sodium salt |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'