missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CREB1 (CREB) Recombinant Human Protein

Expressed from DNA sequence encoding human CREB1 isoform B (Met 1-Asp 327) fused to N-terminal polyhistidine-tagged MBP tag at N-terminus

Brand:  Thermo Scientific™ 10074H10E5

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15244605

  • £63.51 / 5µg


Estimated Shipment
14-06-2018
Add to basket

Description

Description

  • 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween80
Specifications

Specifications

Lyophilized
78.7kd
5μg
−20°C
CAMP responsive element binding protein 1
Assay Standard
E.coli
N-His.MBP Tag
1385
≥90% by SDS-PAGE
Human
Recombinant
CREB1
Transcription Factors
CREB1 (CREB) Recombinant Human Protein
Videos

For Research Use Only. Not for use in diagnostic procedures.