missing translation for 'onlineSavingsMsg'
Learn More

Creatine, anhydrous, 98%, Thermo Scientific Chemicals

Catalog Number p-7023676
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
57-00-1
C4H9N3O2
131.135
MFCD00004282
CVSVTCORWBXHQV-UHFFFAOYSA-N
creatine, creatin, kreatin, krebiozon, n-amidinosarcosine, n-methyl-n-guanylglycine, methylglycocyamine, 2-1-methylguanidino acetic acid, creatine, hydrate, alpha-methylguanido acetic acid
586
CHEBI:16919
2-[carbamimidoyl(methyl)amino]acetic acid
CN(CC(=O)O)C(=N)N
This item is not returnable. View return policy
57-00-1
C4H9N3O2
131.135
MFCD00004282
CVSVTCORWBXHQV-UHFFFAOYSA-N
creatine, creatin, kreatin, krebiozon, n-amidinosarcosine, n-methyl-n-guanylglycine, methylglycocyamine, 2-1-methylguanidino acetic acid, creatine, hydrate, alpha-methylguanido acetic acid
586
CHEBI:16919
2-[carbamimidoyl(methyl)amino]acetic acid
CN(CC(=O)O)C(=N)N

Creatine is involved in the rapid production of ATP associated with creatine kinases in skeletal muscle tissue. Creatine may be used as a supplement to study its mechanism of action. It acts as a nutraceutical and a neuroprotective agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Creatine is involved in the rapid production of ATP associated with creatine kinases in skeletal muscle tissue. Creatine may be used as a supplement to study its mechanism of action. It acts as a nutraceutical and a neuroprotective agent.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY
Melting Point ∼290°C (decomposition)
Density 1.33
Assay Percent Range 98%
Quantity 25 g
Beilstein 907175
Merck Index 14,2568
Solubility Information Soluble in water.
Formula Weight 131.13
Percent Purity 98%
Chemical Name or Material Creatine, anhydrous

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'