missing translation for 'onlineSavingsMsg'
Learn More

Sino Biological™ CRABP2 Recombinant Human Protein, N-His Tag

Expressed from DNA sequence encoding human CRABP2 (Pro2-Glu 138) fused to polyhistide tag at N-terminus

Brand:  Sino Biological™ 11203H07E5

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15288365

  • £63.51 / 5µg


Estimated Shipment
13-03-2018
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

  • Formulation: 20mM Tris, 500mM NaCl, pH 8.0, 5% Mannitol, 5% Trehalose, 0.02% Tween80, 5% Mannitol, 5% Trehalose, 0.02% Tween80
Specifications

Specifications

Lyophilized
16.5kd
5μg
Full Length, Recombinant
Cellular Retinoic Acid Binding Protein 2
Assay Standard
E.coli
His Tag
1382
≥90% by SDS-PAGE
Human
<1 EU/μg
CRABP2
Other proteins
CRABP2 Recombinant Human Protein
Videos