Learn More
CPPG, Tocris Bioscience™
Very potent group III mGlu antagonist
Brand:  Tocris (Bio-Techne) 097250
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13488223
  • £570.72 / 50mg
Estimated Shipment
04-10-2018
Add to basket
Description
Description
Potent group II/III mGlu receptor antagonist, with approximately 20-fold selectivity for group III over group II (IC50 values of 2.2 and 46.2 nM respectively). A much less potent antagonist at group I receptors in neonatal rat cortical slices (KB = 0.65 &plusmin; 0.07 nM). Also available as part of the Group III mGlu Receptor Tocriset™ .
Specifications
Chemical Identifiers
183364-82-1
271.209
(RS)-α-Cyclopropyl-4-phosphonophenylglycine
C11H14NO5P
IGODGTDUQSMDQU-UHFFFAOYSA-N
2878
Specifications
CPPG
>98%
(RS)-α-Cyclopropyl-4-phosphonophenylglycine
C1CC1C(C2=CC=C(C=C2)P(=O)(O)O)(C(=O)O)N
271.209
271.21
50mg
183364-82-1
C11H14NO5P
IGODGTDUQSMDQU-UHFFFAOYSA-N
2-amino-2-cyclopropyl-2-(4-phosphonophenyl)acetic acid
2878
Solid
Videos