missing translation for 'onlineSavingsMsg'
Learn More

CPCCOEt, Tocris Bioscience™

Selective non-competitive mGlu1 antagonist

Brand:  Tocris (Bio-Techne) 1028/10

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13438333

  • £147.15 / 10mg

Estimated Shipment
28-06-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

hmGlu1 subtype-selective non-competitive antagonist (IC50 = 6.5 μM). Has no agonist or antagonist activity at hmGlu2, 4a, 5a, 7b, 8a or ionotropic receptors at concentrations of up to 100 μM.
Specifications

Chemical Identifiers

179067-99-3
247.25
cpccoet
ethyl (1aS,7Z,7aS)-7-hydroxyimino-1,7a-dihydrocyclopropa[b]chromene-1a-carboxylate
C13H13NO4
FXCTZFMSAHZQTR-DRYKPYBASA-N
92460002
CCOC(=O)C12CC1C(=NO)C3=CC=CC=C3O2

Specifications

CPCCOEt
179067-99-3
cpccoet
CCOC(=O)C12CC1C(=NO)C3=CC=CC=C3O2
247.25
247.25
10mg
C13H13NO4
FXCTZFMSAHZQTR-DRYKPYBASA-N
ethyl (1aS,7Z,7aS)-7-hydroxyimino-1,7a-dihydrocyclopropa[b]chromene-1a-carboxylate
92460002
>99%
Videos