missing translation for 'onlineSavingsMsg'
Learn More

CP 99994 dihydrochloride, Tocris Bioscience™

High affinity NK1 antagonist

Brand:  Tocris (Bio-Techne) 341710

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13432984

  • £227.85 / 10mg

Estimated Shipment date
from Supplier 26-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

High affinity NK1 antagonist (Ki = 0.145 nM in vitro). Also displays high ex vivo binding potency in gerbil striatum (IC50 = 36.8 nM). Attenuates endothelium-dependent contraction induced by substance P.
Specifications

Chemical Identifiers

145148-39-6
369.33
2s,3s-1-2-methoxyphenyl methyl-2-phenylpiperidin-3-amine dihydrochloride, 3-piperidinamine,n-2-methoxyphenyl methyl-2-phenyl-,dihydrochloride, 2s,3s
(2S,3S)-1-[(2-methoxyphenyl)methyl]-2-phenylpiperidin-3-amine;dihydrochloride
C19H26Cl2N2O
STUCSMJJXNCIOE-FFUVTKDNSA-N
73013943
COC1=CC=CC=C1CN2CCCC(C2C3=CC=CC=C3)N.Cl.Cl

Specifications

10mg
145148-39-6
2s,3s-1-2-methoxyphenyl methyl-2-phenylpiperidin-3-amine dihydrochloride, 3-piperidinamine,n-2-methoxyphenyl methyl-2-phenyl-,dihydrochloride, 2s,3s
COC1=CC=CC=C1CN2CCCC(C2C3=CC=CC=C3)N.Cl.Cl
369.33
369.33
CP 99994 dihydrochloride
C19H26Cl2N2O
STUCSMJJXNCIOE-FFUVTKDNSA-N
(2S,3S)-1-[(2-methoxyphenyl)methyl]-2-phenylpiperidin-3-amine;dihydrochloride
73013943
>99%
Videos
Documents