missing translation for 'onlineSavingsMsg'
Learn More

CP 945598 hydrochloride, Tocris Bioscience™

CB1 receptor antagonist

Brand:  Tocris (Bio-Techne) 423610

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13464404

  • £245.49 / 10mg

Estimated Shipment date
from Supplier 25-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Selective, high affinity CB1 antagonist (Ki values are 0.7 and 0.12 nM in binding and functional assays respectively). Displays low affinity for CB2 receptors (Ki = 7600 nM).
Specifications

Chemical Identifiers

686347-12-6
546.881
otenabant hydrochloride, cp 945598 hydrochloride, otenabant hcl, cp945598 hydrochloride, otenabant hydrochloride usan, otenabant usan/inn, 1-8-2-chlorophenyl-9-4-chlorophenyl-9h-purin-6-yl-4-ethylamino piperidine-4-carboxamide hydrochloride, 1-8-2-chlorophenyl-9-4-chlorophenyl-9h-purin-6-yl-4-ethylamino piperidine-4-carboxamide monohydrochloride
1-[8-(2-chlorophenyl)-9-(4-chlorophenyl)purin-6-yl]-4-(ethylamino)piperidine-4-carboxamide;hydrochloride
C25H26Cl3N7O
KPYUQCJBZGQHPL-UHFFFAOYSA-N
16223963
CCNC1(CCN(CC1)C2=NC=NC3=C2N=C(N3C4=CC=C(C=C4)Cl)C5=CC=CC=C5Cl)C(=O)N.Cl

Specifications

10mg
686347-12-6
otenabant hydrochloride, cp 945598 hydrochloride, otenabant hcl, cp945598 hydrochloride, otenabant hydrochloride usan, otenabant usan/inn, 1-8-2-chlorophenyl-9-4-chlorophenyl-9h-purin-6-yl-4-ethylamino piperidine-4-carboxamide hydrochloride, 1-8-2-chlorophenyl-9-4-chlorophenyl-9h-purin-6-yl-4-ethylamino piperidine-4-carboxamide monohydrochloride
CCNC1(CCN(CC1)C2=NC=NC3=C2N=C(N3C4=CC=C(C=C4)Cl)C5=CC=CC=C5Cl)C(=O)N.Cl
546.881
546.88
CP 945598 hydrochloride
C25H26Cl3N7O
KPYUQCJBZGQHPL-UHFFFAOYSA-N
1-[8-(2-chlorophenyl)-9-(4-chlorophenyl)purin-6-yl]-4-(ethylamino)piperidine-4-carboxamide;hydrochloride
16223963
>99%
Videos
Documents