missing translation for 'onlineSavingsMsg'
Learn More

CP 94253 hydrochloride, Tocris Bioscience™

Potent and selective 5-HT1B agonist

Brand:  Tocris (Bio-Techne) 131710

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13469123

  • £196.98 / 10mg

Estimated Shipment date
from Supplier 25-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent, selective 5-HT1B agonist (Ki values are 89, 2, 860, 49 and 1,600 nM for 5-HT1A, 5-HT1B, 5-HT1C, 5-HT1D and 5-HT2 receptors respectively). Centrally active upon systemic administration in vivo.
Specifications

Chemical Identifiers

845861-39-4
293.795
cp 94253 hydrochloride, 5-propoxy-3-1,2,3,6-tetrahydro-4-pyridinyl-1h-pyrrolo 3,2-b pyridine hydrochloride, 5-propoxy-3-1,2,3,6-tetrahydropyridin-4-yl-1h-pyrrolo 3,2-b pyridine hydrochloride, zlchem 1209, 5-propoxy-3-1,2,3,6-tetrahydro-4-pyridinyl-1h-pyrrolo 3,2-b pyridinehydrochloride, cp 94253 hydrochloride/, 4-5-propoxy-1h-pyrrolo 3,2-b pyridin-3-yl-1,2,3,6-tetrahydropyridine hydrochloride
5-propoxy-3-(1,2,3,6-tetrahydropyridin-4-yl)-1H-pyrrolo[3,2-b]pyridine;hydrochloride
C15H20ClN3O
PIIOXKQIZCVXMD-UHFFFAOYSA-N
11652258
CCCOC1=NC2=C(C=C1)NC=C2C3=CCNCC3.Cl

Specifications

10mg
845861-39-4
cp 94253 hydrochloride, 5-propoxy-3-1,2,3,6-tetrahydro-4-pyridinyl-1h-pyrrolo 3,2-b pyridine hydrochloride, 5-propoxy-3-1,2,3,6-tetrahydropyridin-4-yl-1h-pyrrolo 3,2-b pyridine hydrochloride, zlchem 1209, 5-propoxy-3-1,2,3,6-tetrahydro-4-pyridinyl-1h-pyrrolo 3,2-b pyridinehydrochloride, cp 94253 hydrochloride/, 4-5-propoxy-1h-pyrrolo 3,2-b pyridin-3-yl-1,2,3,6-tetrahydropyridine hydrochloride
CCCOC1=NC2=C(C=C1)NC=C2C3=CCNCC3.Cl
293.795
293.8
CP 94253 hydrochloride
C15H20ClN3O
PIIOXKQIZCVXMD-UHFFFAOYSA-N
5-propoxy-3-(1,2,3,6-tetrahydropyridin-4-yl)-1H-pyrrolo[3,2-b]pyridine;hydrochloride
11652258
>98%
Videos
Documents