missing translation for 'onlineSavingsMsg'
Learn More

CP 93129 dihydrochloride, Tocris Bioscience™

5-HT1B agonist

Brand:  Tocris (Bio-Techne) 103210

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13488343

  • £151.80 / 10mg


Estimated Shipment
04-10-2018
Add to basket

Description

Description

Potent and highly selective 5-HT1B agonist (Ki values are 8.1, 1100, 1500, 2900 and 7200 nM for 5-HT1B, 5-HT1D, 5-HT1A, 5-HT1c and 5-HT2, respectively). Reduces food intake and decreases body weight in rats.
Specifications

Chemical Identifiers

879089-64-2
306.187
1,4-Dihydro-3-(1,2,3,6-tetrahydro-4-pyridinyl)-5H-pyrrol[3,2-b]pyridin-5-one dihydrochloride
3-(1,2,3,6-tetrahydropyridin-4-yl)-1,4-dihydropyrrolo[3,2-b]pyridin-5-one;hydrate;dihydrochloride
C12H17Cl2N3O2
UXDAADMPPAHNMY-UHFFFAOYSA-N
71311960
C1CNCC=C1C2=CNC3=C2NC(=O)C=C3.O.Cl.Cl

Specifications

CP 93129 dihydrochloride
879089-64-2
C12H17Cl2N3O2
UXDAADMPPAHNMY-UHFFFAOYSA-N
3-(1,2,3,6-tetrahydropyridin-4-yl)-1,4-dihydropyrrolo[3,2-b]pyridin-5-one;hydrate;dihydrochloride
71311960
10mg
>98%
1,4-Dihydro-3-(1,2,3,6-tetrahydro-4-pyridinyl)-5H-pyrrol[3,2-b]pyridin-5-one dihydrochloride
C1CNCC=C1C2=CNC3=C2NC(=O)C=C3.O.Cl.Cl
306.187
288.18
Videos