Learn More
CP 471474, Tocris Bioscience™
Broad spectrum MMP inhibitor
Brand:  Tocris (Bio-Techne) 378050
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13433594
  • £627.69 / 50mg
Estimated Shipment date
from Supplier 26-02-2020
Description
Description
Broad spectrum MMP inhibitor (IC50 values are 0.7, 0.9, 13, 16 and 1170 nM for MMP-2, MMP-13, MMP-9, MMP-3 and MMP-1 respectively). Attenuates early left ventricular dilation after experimental myocardial infarction in mice.
Specifications
Chemical Identifiers
210755-45-6
368.379
2-4-4-fluorophenoxy phenyl sulfonyl amino-n-hydroxy-2-methylpropanamide, cp hplc, 2-4-4-fluorophenoxy phenylsulfonamido-n-hydroxy-2-methylpropanamide, 2-4-4-fluorophenoxy phenyl sulfonyl amino-n-hydrox y-2-methylpropanamide, 2-4-4-fluoro-phenoxy-benzenesulfonylamino-n-hydroxy-2-methyl-propionamide, 2-4-4-fluorophenoxy benzenesulfonamido-n-hydroxy-2-methylpropanamide, propanamide, 2-4-4-fluorophenoxy phenyl sulfonyl amino-n-hydroxy-2-methyl
2-[[4-(4-fluorophenoxy)phenyl]sulfonylamino]-N-hydroxy-2-methylpropanamide
C16H17FN2O5S
QCOQJYRPDUMCNP-UHFFFAOYSA-N
9907286
CC(C)(C(=O)NO)NS(=O)(=O)C1=CC=C(C=C1)OC2=CC=C(C=C2)F
Specifications
50mg
210755-45-6
2-4-4-fluorophenoxy phenyl sulfonyl amino-n-hydroxy-2-methylpropanamide, cp hplc, 2-4-4-fluorophenoxy phenylsulfonamido-n-hydroxy-2-methylpropanamide, 2-4-4-fluorophenoxy phenyl sulfonyl amino-n-hydrox y-2-methylpropanamide, 2-4-4-fluoro-phenoxy-benzenesulfonylamino-n-hydroxy-2-methyl-propionamide, 2-4-4-fluorophenoxy benzenesulfonamido-n-hydroxy-2-methylpropanamide, propanamide, 2-4-4-fluorophenoxy phenyl sulfonyl amino-n-hydroxy-2-methyl
CC(C)(C(=O)NO)NS(=O)(=O)C1=CC=C(C=C1)OC2=CC=C(C=C2)F
368.379
368.38
CP 471474
C16H17FN2O5S
QCOQJYRPDUMCNP-UHFFFAOYSA-N
2-[[4-(4-fluorophenoxy)phenyl]sulfonylamino]-N-hydroxy-2-methylpropanamide
9907286
>97%