missing translation for 'onlineSavingsMsg'
Learn More

CP 465022 hydrochloride, Tocris Bioscience™

Selective, non-competitive AMPA antagonist

Brand:  Tocris (Bio-Techne) 293210

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13462074

  • £155.76 / 10mg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Selective, non-competitive AMPA antagonist (IC50 = 25 nM in rat cortical neurons) that displays potent anticonvulsant activity. Also significantly blocks the persistent component of Nav1.6 channel activity. Brain penetrant and orally active.
Specifications

Chemical Identifiers

199655-36-2
499.411
cp 465022 hydrochloride
3-(2-chlorophenyl)-2-[2-[6-(diethylaminomethyl)pyridin-2-yl]ethenyl]-6-fluoroquinazolin-4-one;hydrochloride
C26H25Cl2FN4O
YKYDGCRJPYLXHY-UHFFFAOYSA-N
67241566
CCN(CC)CC1=CC=CC(=N1)C=CC2=NC3=C(C=C(C=C3)F)C(=O)N2C4=CC=CC=C4Cl.Cl

Specifications

CP 465022 hydrochloride
199655-36-2
cp 465022 hydrochloride
CCN(CC)CC1=CC=CC(=N1)C=CC2=NC3=C(C=C(C=C3)F)C(=O)N2C4=CC=CC=C4Cl.Cl
499.411
499.41
10mg
C26H25Cl2FN4O
YKYDGCRJPYLXHY-UHFFFAOYSA-N
3-(2-chlorophenyl)-2-[2-[6-(diethylaminomethyl)pyridin-2-yl]ethenyl]-6-fluoroquinazolin-4-one;hydrochloride
67241566
>99%
Videos